missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(Anilinomethyl)pyrrolidine, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 445800010

 View more versions of this product

Catalog No. AC445800010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

(S)-(+)-2-(Anilinomethyl)pyrrolidine
126°C (0.1 kPa)
C11H16N2
+ 1.63
MCHWKJRTMPIHRA-NSHDSACASA-N
N-[[(2S)-pyrrolidin-2-yl]methyl]aniline
6950385
95%
64030-44-0
95%
1g
s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline
C1CC(NC1)CNC2=CC=CC=C2
176.26
176.26

Chemical Identifiers

64030-44-0
176.26
s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline
N-[[(2S)-pyrrolidin-2-yl]methyl]aniline
C11H16N2
MCHWKJRTMPIHRA-NSHDSACASA-N
6950385
C1CC(NC1)CNC2=CC=CC=C2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only