missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(Anilinomethyl)pyrrolidine, 95%, Thermo Scientific™

Chemical Identifiers

CAS 64030-44-0
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
InChI Key MCHWKJRTMPIHRA-NSHDSACASA-N
Synonym s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline
PubChem CID 6950385
IUPAC Name N-[[(2S)-pyrrolidin-2-yl]methyl]aniline
SMILES C1CC(NC1)CNC2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445800010
SDS View Documents
Thermo Scientific Chemicals
445800010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

64030-44-0
176.26
s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline
N-[[(2S)-pyrrolidin-2-yl]methyl]aniline
C11H16N2
MCHWKJRTMPIHRA-NSHDSACASA-N
6950385
C1CC(NC1)CNC2=CC=CC=C2

Specifications

64030-44-0
95%
1g
s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline
C1CC(NC1)CNC2=CC=CC=C2
176.26
176.26
(S)-(+)-2-(Anilinomethyl)pyrrolidine
126°C (0.1 kPa)
C11H16N2
+ 1.63
MCHWKJRTMPIHRA-NSHDSACASA-N
N-[[(2S)-pyrrolidin-2-yl]methyl]aniline
6950385
95%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only