missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine 98.0+%, TCI America™

Catalog No. A24731G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24731G Supplier TCI America Supplier No. A24731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 119020-01-8
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
MDL Number MFCD03419257
InChI Key SOGXYCNKQQJEED-QMMMGPOBSA-N
Synonym s-1-boc-2-aminomethyl pyrrolidine, s-1-n-boc-2-aminomethyl pyrrolidine, s-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-aminomethyl pyrrolidine-1-carboxylate, s-2-aminomethyl-1-n-boc-pyrrolidine, s-2-aminomethyl-1-boc-pyrrolidine, tert-butyl s-2-aminomethylpyrrolidine-1-carboxylate, s-2-aminomethyl-1-n-boc-pyyrolidine
PubChem CID 1512533
IUPAC Name tert-butyl (2S)-2-(aminomethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC[C@H]1CN

Specifications

Color Yellow
Boiling Point 112°C
Quantity 1 g
UN Number 2735
Formula Weight 200.28
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine
TSCA No
Recommended Storage Refrigerator