accessibility menu, dialog, popup

UserName

CH3H3CCH3OON(S)H2N

CAS RN 119020-01-8

IUPAC Name: tert-butyl (2S)-2-(aminomethyl)pyrrolidine-1-carboxylate
Synonyms: tert-butyl (2S)-2-(aminomethyl)pyrrolidine-1-carboxylate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: SOGXYCNKQQJEED-QMMMGPOBSA-N
SMILES: CC(C)(C)OC(=O)N1CCC[C@H]1CN
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)