missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 98.0+%, TCI America™
$233.00 - $848.14
Chemical Identifiers
| CAS | 22795-99-9 |
|---|---|
| Molecular Formula | C7H16N2 |
| Molecular Weight (g/mol) | 128.219 |
| MDL Number | MFCD00191371 |
| InChI Key | UNRBEYYLYRXYCG-ZETCQYMHSA-N |
| Synonym | s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590 |
| PubChem CID | 643457 |
| IUPAC Name | [(2S)-1-ethylpyrrolidin-2-yl]methanamine |
| SMILES | CCN1CCCC1CN |
Chemical Identifiers
| 22795-99-9 | |
| 128.219 | |
| UNRBEYYLYRXYCG-ZETCQYMHSA-N | |
| 643457 | |
| CCN1CCCC1CN |
| C7H16N2 | |
| MFCD00191371 | |
| s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590 | |
| [(2S)-1-ethylpyrrolidin-2-yl]methanamine |
Specifications
| 22795-99-9 | |
| C7H16N2 | |
| 1 g | |
| s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590 | |
| CCN1CCCC1CN | |
| 128.219 | |
| 128.22 | |
| Liquid |
| Yellow | |
| MFCD00191371 | |
| 2733 | |
| UNRBEYYLYRXYCG-ZETCQYMHSA-N | |
| [(2S)-1-ethylpyrrolidin-2-yl]methanamine | |
| 643457 | |
| ≥98.0% (GC,T) | |
| (S)-(-)-2-Aminomethyl-1-ethylpyrrolidine |
Safety and Handling
EINECSNumber : (9)-0211
TSCA : No