missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 98.0+%, TCI America™
Supplier: TCI America A13015G
Specifications
| (S)-(-)-2-Aminomethyl-1-ethylpyrrolidine | |
| Yellow | |
| MFCD00191371 | |
| 2733 | |
| UNRBEYYLYRXYCG-ZETCQYMHSA-N | |
| [(2S)-1-ethylpyrrolidin-2-yl]methanamine | |
| 643457 | |
| ≥98.0% (GC,T) |
| 22795-99-9 | |
| C7H16N2 | |
| 5 g | |
| s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590 | |
| CCN1CCCC1CN | |
| 128.219 | |
| 128.22 | |
| Liquid |
Chemical Identifiers
| 22795-99-9 | |
| 128.219 | |
| UNRBEYYLYRXYCG-ZETCQYMHSA-N | |
| 643457 | |
| CCN1CCCC1CN |
| C7H16N2 | |
| MFCD00191371 | |
| s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590 | |
| [(2S)-1-ethylpyrrolidin-2-yl]methanamine |
Safety and Handling
EINECSNumber : (9)-0211
TSCA : No