Learn More
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine, 95%
CAS: 22795-99-9 | C7H16N2 | 128.219 g/mol
$99.58 - $323.05
Chemical Identifiers
| CAS | 22795-99-9 |
|---|---|
| Molecular Formula | C7H16N2 |
| Molecular Weight (g/mol) | 128.219 |
| MDL Number | MFCD00191371 |
| InChI Key | UNRBEYYLYRXYCG-ZETCQYMHSA-N |
| Synonym | s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590 |
| PubChem CID | 643457 |
| IUPAC Name | [(2S)-1-ethylpyrrolidin-2-yl]methanamine |
| SMILES | CCN1CCCC1CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5713703
|
Thermo Scientific Chemicals
H5713703 |
1 g |
Each for $99.58
|
|
|||||
|
AAH5713706
|
Thermo Scientific Chemicals
H5713706 |
5 g |
Each for $323.05
|
|
|||||
Description
It is used as a pharmaceutical intermediate of antipsychotics levosulpiride and for the preparation of medicines and other fine chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate of antipsychotics levosulpiride and for the preparation of medicines and other fine chemicals.
Solubility
Soluble in chloroform, methanol. Slightly soluble in water.
Notes
Hygroscopic. Store away from acids, water/moisture, acid chlorides, acid anhydrides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Chemical Identifiers
| 22795-99-9 | |
| 128.219 | |
| UNRBEYYLYRXYCG-ZETCQYMHSA-N | |
| 643457 | |
| CCN1CCCC1CN |
| C7H16N2 | |
| MFCD00191371 | |
| s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590 | |
| [(2S)-1-ethylpyrrolidin-2-yl]methanamine |
Specifications
| 22795-99-9 | |
| 50°C to 54°C | |
| C7H16N2 | |
| MFCD00191371 | |
| UN2733 | |
| s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590 | |
| UNRBEYYLYRXYCG-ZETCQYMHSA-N | |
| CCN1CCCC1CN | |
| 128.219 | |
| 128.22 | |
| (S)-(-)-2-Aminomethyl-1-ethylpyrrolidine |
| 0.919 | |
| 57°C (135°F) | |
| 1.466 | |
| 1 g | |
| Hygroscopic | |
| Soluble in chloroform,methanol. Slightly soluble in water. | |
| −95° (c=1 in Methanol) | |
| [(2S)-1-ethylpyrrolidin-2-yl]methanamine | |
| 643457 | |
| 95% |
Safety and Handling
GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302+H312-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: I
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only