missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine, 95%

CAS: 22795-99-9 | C7H16N2 | 128.219 g/mol

Supplier:  Thermo Scientific Chemicals H5713706

 View more versions of this product

Catalog No. AAH5713706


Only null left
Explore available promotions

Description

Description

It is used as a pharmaceutical intermediate of antipsychotics levosulpiride and for the preparation of medicines and other fine chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate of antipsychotics levosulpiride and for the preparation of medicines and other fine chemicals.

Solubility
Soluble in chloroform, methanol. Slightly soluble in water.

Notes
Hygroscopic. Store away from acids, water/moisture, acid chlorides, acid anhydrides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Specifications

Specifications

(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine
0.919
57°C (135°F)
1.466
5 g
Hygroscopic
Soluble in chloroform,methanol. Slightly soluble in water.
−95° (c=1 in Methanol)
[(2S)-1-ethylpyrrolidin-2-yl]methanamine
643457
95%
22795-99-9
50°C to 54°C
C7H16N2
MFCD00191371
UN2733
s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590
UNRBEYYLYRXYCG-ZETCQYMHSA-N
CCN1CCCC1CN
128.219
128.22

Chemical Identifiers

22795-99-9
128.219
UNRBEYYLYRXYCG-ZETCQYMHSA-N
643457
CCN1CCCC1CN
C7H16N2
MFCD00191371
s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590
[(2S)-1-ethylpyrrolidin-2-yl]methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302+H312-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: I

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only