missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Aminomethyl-1-Boc-pyrrolidine, 97%

Catalog No. p-7035025
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5272703 1 g
AAH5272706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5272703 Supplier Thermo Scientific Chemicals Supplier No. H5272703

CAS: 119020-01-8 | C10H20N2O2 | 200.28 g/mol

(S)-2-Aminomethyl-1-Boc-pyrrolidine acts as an intermediate in organic synthesis. It is also used as a chiral and heterocyclic building block in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-2-Aminomethyl-1-Boc-pyrrolidine acts as an intermediate in organic synthesis. It is also used as a chiral and heterocyclic building block in synthetic chemistry.

Solubility
Miscible with N-methylpyrrolidinone.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 119020-01-8
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
MDL Number MFCD03419257
InChI Key SOGXYCNKQQJEED-QMMMGPOBSA-N
Synonym s-1-boc-2-aminomethyl pyrrolidine, s-1-n-boc-2-aminomethyl pyrrolidine, s-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-aminomethyl pyrrolidine-1-carboxylate, s-2-aminomethyl-1-n-boc-pyrrolidine, s-2-aminomethyl-1-boc-pyrrolidine, tert-butyl s-2-aminomethylpyrrolidine-1-carboxylate, s-2-aminomethyl-1-n-boc-pyyrolidine
PubChem CID 1512533
IUPAC Name tert-butyl (2S)-2-(aminomethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC[C@H]1CN

Specifications

Boiling Point 98°C to 112°C (1 mmHg)
Quantity 1 g
UN Number UN2735
Solubility Information Miscible with N-methylpyrrolidinone.
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (S)-2-Aminomethyl-1-Boc-pyrrolidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only