missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminobutyric Acid 99.0+%, TCI America™

Catalog No. A08261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08261G 1 g
A08265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08261G Supplier TCI America Supplier No. A08261G
Only null left

Chemical Identifiers

CAS 1492-24-6
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00064415
InChI Key QWCKQJZIFLGMSD-VKHMYHEASA-N
Synonym l-2-aminobutyric acid, s-2-aminobutanoic acid, 2s-2-aminobutanoic acid, h-abu-oh, l-alpha-aminobutyric acid, s-+-2-aminobutyric acid, l-+-2-aminobutyric acid, --2-aminobutyric acid, s-2-aminobutyric acid, s-2-amino-butyric acid
PubChem CID 80283
ChEBI CHEBI:35619
IUPAC Name (2S)-2-aminobutanoic acid
SMILES CCC(C(=O)O)N

Specifications

Melting Point 280°C
Color White
Quantity 1 g
Formula Weight 103.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-Aminobutyric Acid
EINECSNumber (2)-2482
TSCA Yes