missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-Aminobutyramide Hydrochloride 98.0+%, TCI America™
Supplier: TCI America A225225G
Specifications
| (S)-2-Aminobutyramide Hydrochloride | |
| 263°C | |
| C4H11ClN2O | |
| 25 g | |
| HDBMIDJFXOYCGK-DFWYDOINSA-N | |
| (2S)-2-aminobutanamide;hydrochloride | |
| 11961924 | |
| ≥98.0% (HPLC,N) |
| 7682-20-4 | |
| White | |
| MFCD00136565 | |
| s-2-aminobutanamide hydrochloride, s-2-aminobutyramide hydrochloride, h-abu-nh2 hcl, 2s-2-aminobutanamide hydrochloride, l-2-aminobutanamide hydrochloride, l-homoalanylamide hcl, s-+-2-aminobutanamide hydrochloride, butanamide, 2-amino-, monohydrochloride, 2s, s-2-aminobutanamide hcl, s-2-amino butanamide hcl | |
| CCC(C(=O)N)N.Cl | |
| 138.595 | |
| 138.60 | |
| Crystalline Powder |
Chemical Identifiers
| 7682-20-4 | |
| 138.595 | |
| HDBMIDJFXOYCGK-DFWYDOINSA-N | |
| 11961924 | |
| CCC(C(=O)N)N.Cl |
| C4H11ClN2O | |
| MFCD00136565 | |
| s-2-aminobutanamide hydrochloride, s-2-aminobutyramide hydrochloride, h-abu-nh2 hcl, 2s-2-aminobutanamide hydrochloride, l-2-aminobutanamide hydrochloride, l-homoalanylamide hcl, s-+-2-aminobutanamide hydrochloride, butanamide, 2-amino-, monohydrochloride, 2s, s-2-aminobutanamide hcl, s-2-amino butanamide hcl | |
| (2S)-2-aminobutanamide;hydrochloride |
Safety and Handling
TSCA : No