missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Aminobutyramide Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117803
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22525G 5 g
A225225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22525G Supplier TCI America Supplier No. A22525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7682-20-4
Molecular Formula C4H11ClN2O
Molecular Weight (g/mol) 138.595
MDL Number MFCD00136565
InChI Key HDBMIDJFXOYCGK-DFWYDOINSA-N
Synonym s-2-aminobutanamide hydrochloride, s-2-aminobutyramide hydrochloride, h-abu-nh2 hcl, 2s-2-aminobutanamide hydrochloride, l-2-aminobutanamide hydrochloride, l-homoalanylamide hcl, s-+-2-aminobutanamide hydrochloride, butanamide, 2-amino-, monohydrochloride, 2s, s-2-aminobutanamide hcl, s-2-amino butanamide hcl
PubChem CID 11961924
IUPAC Name (2S)-2-aminobutanamide;hydrochloride
SMILES CCC(C(=O)N)N.Cl

Specifications

Melting Point 263°C
Color White
Quantity 5 g
Formula Weight 138.60
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-Aminobutyramide Hydrochloride