missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminobutane, 98%

CAS: 513-49-5 | C4H11N | 73.139 g/mol

Supplier:  Thermo Scientific Chemicals L1006906

 View more versions of this product

Catalog No. AAL1006906


Only null left
Explore available promotions

Description

Description

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

(S)-(+)-2-Aminobutane
0.722
63°C
Amine-like
1.393
5 g
1718760
14,1544
Fully miscible in water.
+7.5° (Neat)
(2S)-butan-2-amine
6713753
98%
513-49-5
12.5
−19°C (−2°F)
C4H11N
MFCD00064417
UN2733
Air Sensitive
s-+-2-aminobutane, s-+-sec-butylamine, 2s-butan-2-amine, 2-butanamine, 2s, s-2-butylamine, s-sec-butylamine, +-2-butylamine, unii-z192xwh21o, 2-butanamine, s
BHRZNVHARXXAHW-BYPYZUCNSA-N
CCC(C)N
73.139
73.14

Chemical Identifiers

513-49-5
73.139
BHRZNVHARXXAHW-BYPYZUCNSA-N
6713753
CCC(C)N
C4H11N
MFCD00064417
s-+-2-aminobutane, s-+-sec-butylamine, 2s-butan-2-amine, 2-butanamine, 2s, s-2-butylamine, s-sec-butylamine, +-2-butylamine, unii-z192xwh21o, 2-butanamine, s
(2S)-butan-2-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318-H302-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H332-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

EINECSNumber : 208-164-7

RTECSNumber : EO3327000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only