missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminobutane, 98%

Catalog No. p-7045266
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL10069MD 250 mg
AAL1006903 1 g
AAL1006906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL10069MD Supplier Thermo Scientific Chemicals Supplier No. L10069MD
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 513-49-5 | C4H11N | 73.139 g/mol

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 513-49-5
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00064417
InChI Key BHRZNVHARXXAHW-BYPYZUCNSA-N
Synonym s-+-2-aminobutane, s-+-sec-butylamine, 2s-butan-2-amine, 2-butanamine, 2s, s-2-butylamine, s-sec-butylamine, +-2-butylamine, unii-z192xwh21o, 2-butanamine, s
PubChem CID 6713753
IUPAC Name (2S)-butan-2-amine
SMILES CCC(C)N

Specifications

Density 0.722
pH 12.5
Boiling Point 63°C
Flash Point −19°C (−2°F)
Odor Amine-like
Refractive Index 1.393
Quantity 250 mg
UN Number UN2733
Beilstein 1718760
Sensitivity Air Sensitive
Merck Index 14,1544
Solubility Information Fully miscible in water.
Optical Rotation +7.5° (Neat)
Formula Weight 73.14
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Aminobutane
Show More Show Less
Hazard Category H225-H302+H332-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318-H302-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 208-164-7
RTECSNumber EO3327000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only