Learn More
(S)-(+)-2-Amino-4-phosphonobutyric acid, 97%, Thermo Scientific Chemicals
A NMDA agonist
Supplier: Thermo Scientific Chemicals J64075MA
Description
A NMDA agonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (S)-(+)-2-Amino-4-phosphonobutyric acid | |
| White | |
| MFCD00083244 | |
| Light sensitive | |
| Soluble to 5 mM in water and to 100 mM in 1eq. NaOH | |
| +24° (c=1 in 6N HCl) | |
| (2S)-2-amino-4-phosphonobutanoic acid | |
| 179394 | |
| 99% |
| 23052-81-5 | |
| C4H10NO5P | |
| 10 mg | |
| l-ap4, 2s-2-amino-4-phosphonobutanoic acid, l-+-2-amino-4-phosphonobutyric acid, chembl33567, s-2-amino-4-phosphonobutyrate, l-ap-4, 3h ap4, l-2-amino-4-phosphonobutiric acid, l-1-amino-4-phosphonobutanoic acid, s-2-amino-4-phosphonobutanoic acid | |
| DDOQBQRIEWHWBT-VKHMYHEASA-N | |
| C(CP(=O)(O)O)C(C(=O)O)N | |
| 183.1 | |
| 183.1 | |
| Powder |
Chemical Identifiers
| 23052-81-5 | |
| 183.1 | |
| DDOQBQRIEWHWBT-VKHMYHEASA-N | |
| 179394 | |
| C(CP(=O)(O)O)C(C(=O)O)N |
| C4H10NO5P | |
| MFCD00083244 | |
| l-ap4, 2s-2-amino-4-phosphonobutanoic acid, l-+-2-amino-4-phosphonobutyric acid, chembl33567, s-2-amino-4-phosphonobutyrate, l-ap-4, 3h ap4, l-2-amino-4-phosphonobutiric acid, l-1-amino-4-phosphonobutanoic acid, s-2-amino-4-phosphonobutanoic acid | |
| (2S)-2-amino-4-phosphonobutanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : EK7713874
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only