accessibility menu, dialog, popup

UserName

CAS RN 23052-81-5

IUPAC Name: (2S)-2-azaniumyl-4-phosphonatobutanoate
Synonyms: L-AP4
Molecular Weight (g/mol): 181.09
Molecular Formula: C4H8NO5P
InChi Key: DDOQBQRIEWHWBT-VKHMYHEASA-L
SMILES: [NH3+][C@@H](CCP([O-])([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results