missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-4-bromobutyric Acid Hydrobromide 98.0+%, TCI America™

Catalog No. A14505G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14505G 5 g
A14501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14505G Supplier TCI America Supplier No. A14505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15159-65-6
Molecular Formula C4H8BrNO2
Molecular Weight (g/mol) 182.02
MDL Number MFCD01631290
InChI Key LMYNKFDVIZBWDF-VKHMYHEASA-N
Synonym s-+-2-amino-4-bromobutyric acid hydrobromide, s-2-amino-4-bromobutanoic acid hydrobromide, l +-2-amino-4-bromobutyric acid hydrobromide, 2s-2-amino-4-bromobutanoic acid hydrobromide, s-+-2-amino-4-bromobutyric acid hbr, pubchem13813, 2s-2-amino-4-bromo-butanoic acid hydrobromide, s-2-amino-4-bromobutanoic acid hbr, s-2-amino-4-bromobutyric acid hydrobromide, s-2-amino-4-bromo-butyric acid hydrobromide
PubChem CID 2733672
IUPAC Name (2S)-2-azaniumyl-4-bromobutanoate
SMILES [NH3+][C@@H](CCBr)C([O-])=O

Specifications

Melting Point 184°C
Quantity 5 g
Formula Weight 262.93
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-Amino-4-bromobutyric Acid Hydrobromide