missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Amino-3-benzyloxy-1-propanol hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. AAH5229703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5229703 1 g
AAH5229706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5229703 Supplier Thermo Scientific Chemicals Supplier No. H5229703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61366-43-6 | Molecular Formula: C10H16ClNO2 | Molecular Weight: 217.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61366-43-6
Molecular Formula C10H16ClNO2
Molecular Weight (g/mol) 217.69
MDL Number MFCD07368914
InChI Key BJWHQBTUHVIRJJ-UHFFFAOYNA-N
Synonym s-2-amino-3-benzyloxy propan-1-ol hydrochloride, s-2-amino-3-benzyloxy-1-propanol hydrochloride, 2s-2-amino-3-benzyloxy propan-1-ol hydrochloride, s---2-amino-3-benzyloxy-1-propanol??hcl, s-2-amino-3-benzyloxy-propan-1-ol hydrochloride, 2s-2-amino-3-benzyloxy propan-1-ol-hydrogen chloride 1/1
PubChem CID 53338757
SMILES Cl.NC(CO)COCC1=CC=CC=C1

Specifications

Quantity 1 g
IUPAC Name (2S)-2-amino-3-phenylmethoxypropan-1-ol;hydrochloride
Formula Weight 217.7
Percent Purity ≥98%
Chemical Name or Material (S)-2-Amino-3-benzyloxy-1-propanol hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only