missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Amino-3-benzyloxy-1-propanol, 98+%

Catalog No. AAH5230403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5230403 1 g
AAH5230406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5230403 Supplier Thermo Scientific Chemicals Supplier No. H5230403
Only null left

CAS: 58577-88-1 | C10H15NO2 | 181.235 g/mol

(S)-(-)-2-Amino-3-benzyloxy-1-propanol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-2-Amino-3-benzyloxy-1-propanol is used as an organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.

Chemical Identifiers

CAS 58577-88-1
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.235
MDL Number MFCD04112477
InChI Key ZJUOMDNENVWMPL-JTQLQIEISA-N
Synonym s-2-amino-3-benzyloxy-1-propanol, h-d-ser bzl-ol, 2s-2-amino-3-benzyloxy propan-1-ol, s-2-amino-3-benzyloxy propan-1-ol, 1-propanol, 2-amino-3-phenylmethoxy-, 2s, s---2-amino-3-benzyloxy-1-propanol, 98+%, ambotzhal1035, s---2-amino-3-benzyloxy-1-propal, s---2-amino-3-benzyloxy-1-propanol
PubChem CID 11401131
IUPAC Name (2S)-2-amino-3-phenylmethoxypropan-1-ol
SMILES C1=CC=C(C=C1)COCC(CO)N

Specifications

Quantity 1 g
UN Number UN3267
Sensitivity Air Sensitive
Formula Weight 181.23
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-2-Amino-3-benzyloxy-1-propanol
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only