missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Amino-2-methyl-4-pentenoic acid, 98%, ee 99+%

Catalog No. p-7036604
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54922MD 250 mg
AAH5492203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54922MD Supplier Thermo Scientific Chemicals Supplier No. H54922MD
Only null left

CAS: 96886-55-4 | C6H11NO2 | 129.159 g/mol

It is used in organic synthesis and as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic synthesis and as an pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Store at 4°C. Keep away from oxidizing agents and heat.

Chemical Identifiers

CAS 96886-55-4
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00145248
InChI Key QMBTZYHBJFPEJB-LURJTMIESA-N
Synonym s-2-amino-2-methylpent-4-enoic acid, s-2-amino-2-methyl-4-pentenoic acid, l-alpha-allylalanine, 2s-2-amino-2-methylpent-4-enoic acid, alpha-allyl-l-ala, r-2-amino-2-methyl-4-pentenoic acid, alpha-methyl-l-allylglycine, s---alpha-allylalanine, 4-pentenoic acid, 2-amino-2-methyl-, 2s, s---2-amino-2-methyl-4-pentenoic acid
PubChem CID 2724753
IUPAC Name (2S)-2-amino-2-methylpent-4-enoic acid
SMILES CC(CC=C)(C(=O)O)N

Specifications

Melting Point 285°C
Quantity 250 mg
Solubility Information Very soluble in water.
Optical Rotation −14° (c=1.3 in 1N HCl)
Formula Weight 129.16
Percent Purity 98%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-2-Amino-2-methyl-4-pentenoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only