missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-1-propanol 98.0+%, TCI America™

Catalog No. p-7117234
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10851ML 1 mL
A10855ML 5 mL
A108525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A10851ML Supplier TCI America Supplier No. A10851ML
Only null left

Chemical Identifiers

CAS 2749-11-3
Molecular Formula C3H10NO
Molecular Weight (g/mol) 76.12
MDL Number MFCD00064412
InChI Key BKMMTJMQCTUHRP-VKHMYHEASA-O
Synonym l-alaninol, s-+-2-amino-1-propanol, s-2-aminopropan-1-ol, 2s-2-aminopropan-1-ol, h-alaninol, alaninol, s-2-amino-1-propanol, +-2-aminopropan-1-ol, 2-aminopropanol, +, unii-v403gh89l1
PubChem CID 80307
ChEBI CHEBI:78502
IUPAC Name (2S)-2-aminopropan-1-ol
SMILES CC(CO)N

Specifications

Color Yellow
Boiling Point 98°C
Quantity 1 mL
UN Number 2735
Formula Weight 75.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Amino-1-propanol
EINECSNumber (9)-0122&(2)-0323
TSCA No