accessibility menu, dialog, popup

UserName

CH3(S)H3NHO

CAS RN 2749-11-3

IUPAC Name: (2S)-1-hydroxypropan-2-aminium
Synonyms: (2S)-1-hydroxypropan-2-aminium
Molecular Weight (g/mol): 76.12
Molecular Formula: C3H10NO
InChi Key: BKMMTJMQCTUHRP-VKHMYHEASA-O
SMILES: C[C@H]([NH3+])CO
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)