missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Amino-1-propanol, 98%

Catalog No. AAB2491606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2491606 5 g
AAB2491614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2491606 Supplier Thermo Scientific Chemicals Supplier No. B2491606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2749-11-3 | C3H10NO | 76.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2749-11-3
MDL Number MFCD00064412
ChEBI CHEBI:78502

Specifications

Density 0.965
Boiling Point 174°C to 176°C
Flash Point 83°C (181°F)
Molecular Formula C3H10NO
Refractive Index 1.4498
Quantity 5 g
UN Number UN2735
Beilstein 1718865
Sensitivity Air sensitive; Hygroscopic
Synonym l-alaninol, s-+-2-amino-1-propanol, s-2-aminopropan-1-ol, 2s-2-aminopropan-1-ol, h-alaninol, alaninol, s-2-amino-1-propanol, +-2-aminopropan-1-ol, 2-aminopropanol, +, unii-v403gh89l1
InChI Key BKMMTJMQCTUHRP-VKHMYHEASA-O
Optical Rotation +22° (c=2 in Ethanol)
SMILES CC(CO)N
IUPAC Name (2S)-2-aminopropan-1-ol
Molecular Weight (g/mol) 76.12
PubChem CID 80307
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Amino-1-propanol
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 220-388-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only