missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole 98.0+%, TCI America™
Supplier: TCI America D43371G
Specifications
| (S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole | |
| 229°C | |
| C7H11N3S | |
| 1 g | |
| DRRYZHHKWSHHFT-BYPYZUCNSA-N | |
| (6S)-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine | |
| 11521153 | |
| ≥98.0% (GC,T) |
| 106092-09-5 | |
| White-Yellow | |
| MFCD07368003 | |
| (S)-N-Despropyl Pramipexole | |
| C1CC2=C(CC1N)SC(=N2)N | |
| 169.246 | |
| 169.25 | |
| Crystalline Powder |
Chemical Identifiers
| 106092-09-5 | |
| 169.246 | |
| DRRYZHHKWSHHFT-BYPYZUCNSA-N | |
| 11521153 | |
| C1CC2=C(CC1N)SC(=N2)N |
| C7H11N3S | |
| MFCD07368003 | |
| (S)-N-Despropyl Pramipexole | |
| (6S)-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator