missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole 98.0+%, TCI America™

Catalog No. p-7128231
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43371G 1 g
D43375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43371G Supplier TCI America Supplier No. D43371G

Chemical Identifiers

CAS 106092-09-5
Molecular Formula C7H11N3S
Molecular Weight (g/mol) 169.246
MDL Number MFCD07368003
InChI Key DRRYZHHKWSHHFT-BYPYZUCNSA-N
Synonym (S)-N-Despropyl Pramipexole
PubChem CID 11521153
IUPAC Name (6S)-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
SMILES C1CC2=C(CC1N)SC(=N2)N

Specifications

Melting Point 229°C
Color White-Yellow
Quantity 1 g
Formula Weight 169.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole
TSCA No
Recommended Storage Refrigerator