missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(4-Isobutylphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. I05491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05491G 1 g
I05495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05491G Supplier TCI America Supplier No. I05491G
Only null left

Chemical Identifiers

CAS 51146-56-6
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00069289
InChI Key HEFNNWSXXWATRW-JTQLQIEISA-N
Synonym s-+-ibuprofen, dexibuprofen, s +-ibuprofen, s-2-4-isobutylphenyl propanoic acid, s-ibuprofen, d-ibuproten, s-+-2-4-isobutylphenyl propionic acid, s-+-4-isobutyl-alpha-methylphenylacetic acid, +-s-p-isobutylhydratropic acid, seractil
PubChem CID 39912
ChEBI CHEBI:43415
IUPAC Name (2S)-2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(=O)O

Specifications

Melting Point 52°C
Color White
Quantity 1 g
Formula Weight 206.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-(4-Isobutylphenyl)propionic Acid