missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-Toluenesulfonate 98.0+%, TCI America™
$369.08 - $1202.32
Chemical Identifiers
| CAS | 23735-43-5 |
|---|---|
| Molecular Formula | C13H18O5S |
| Molecular Weight (g/mol) | 286.342 |
| MDL Number | MFCD00063234 |
| InChI Key | SRKDUHUULIWXFT-NSHDSACASA-N |
| Synonym | s-+-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate, s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, s-+-2,2-dimethyl-1,3-dioxolane-4-ylmethyl p-toluenesulfonate, s-2,2-dimethyl-1,3-dioxolane-4-methanol p-toluenesulfonate, r---2,2-dimethyl-1,3-dioxolane-4-ylmethyl-p-toluenesulfonate, pubchem6795, 2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate #, s-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate |
| PubChem CID | 2723760 |
| IUPAC Name | [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)OCC2COC(O2)(C)C |
Chemical Identifiers
| 23735-43-5 | |
| 286.342 | |
| SRKDUHUULIWXFT-NSHDSACASA-N | |
| 2723760 | |
| CC1=CC=C(C=C1)S(=O)(=O)OCC2COC(O2)(C)C |
| C13H18O5S | |
| MFCD00063234 | |
| s-+-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate, s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, s-+-2,2-dimethyl-1,3-dioxolane-4-ylmethyl p-toluenesulfonate, s-2,2-dimethyl-1,3-dioxolane-4-methanol p-toluenesulfonate, r---2,2-dimethyl-1,3-dioxolane-4-ylmethyl-p-toluenesulfonate, pubchem6795, 2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate #, s-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate | |
| [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate |
Specifications
| 23735-43-5 | |
| C13H18O5S | |
| 1 g | |
| SRKDUHUULIWXFT-NSHDSACASA-N | |
| [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate | |
| 2723760 | |
| ≥98.0% (GC) | |
| (S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-Toluenesulfonate |
| White-Yellow | |
| MFCD00063234 | |
| s-+-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate, s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, s-+-2,2-dimethyl-1,3-dioxolane-4-ylmethyl p-toluenesulfonate, s-2,2-dimethyl-1,3-dioxolane-4-methanol p-toluenesulfonate, r---2,2-dimethyl-1,3-dioxolane-4-ylmethyl-p-toluenesulfonate, pubchem6795, 2,2-dimethyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate #, s-2,2-dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate | |
| CC1=CC=C(C=C1)S(=O)(=O)OCC2COC(O2)(C)C | |
| 286.342 | |
| 286.34 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator