missing translation for 'onlineSavingsMsg'
Learn More

(S)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl 98.0+%, TCI America™

Catalog No. B32035G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32035G 5 g
B32031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32035G Supplier TCI America Supplier No. B32035G

Chemical Identifiers

CAS 142128-92-5
Molecular Formula C24H22O4
Molecular Weight (g/mol) 374.44
MDL Number MFCD03788935
InChI Key YIAQRNNJNMLGTP-UHFFFAOYSA-N
Synonym r-2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthyl, 2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-bis methoxymethoxy, s---2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-2,2/'-bis methoxymethoxy-1,1/'-binaphthalene
PubChem CID 10959708
IUPAC Name 2,2'-bis(methoxymethoxy)-1,1'-binaphthalene
SMILES COCOC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OCOC

Specifications

Melting Point 102°C
Color White
Quantity 5 g
Formula Weight 374.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2,2′-Bis(methoxymethoxy)-1,1′-binaphthyl