accessibility menu, dialog, popup

UserName

CH3OOOOCH3(?)

CAS RN 142128-92-5

IUPAC Name: 2,2'-bis(methoxymethoxy)-1,1'-binaphthalene
Synonyms: 2,2'-bis(methoxymethoxy)-1,1'-binaphthalene
Molecular Weight (g/mol): 374.44
Molecular Formula: C24H22O4
InChi Key: YIAQRNNJNMLGTP-UHFFFAOYSA-N
SMILES: COCOC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OCOC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results