missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific™
Chemical Identifiers
| CAS | 1365531-82-3 |
|---|---|
| Molecular Formula | C44H44O4P2 |
| Molecular Weight (g/mol) | 698.78 |
| MDL Number | MFCD19443623 |
| InChI Key | JXFDRZQHVPTIMH-UHFFFAOYSA-N |
| Synonym | s-2,2'-bis di-p-tolylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine |
| PubChem CID | 87110704 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH6092103
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1365531-82-3 | |
| 698.78 | |
| JXFDRZQHVPTIMH-UHFFFAOYSA-N | |
| 87110704 |
| C44H44O4P2 | |
| MFCD19443623 | |
| s-2,2'-bis di-p-tolylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine |
Specifications
| 1365531-82-3 | |
| MFCD19443623 | |
| s-2,2'-bis di-p-tolylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine | |
| 698.78 | |
| 698.77 | |
| (S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl |
| C44H44O4P2 | |
| Air sensitive | |
| JXFDRZQHVPTIMH-UHFFFAOYSA-N | |
| 87110704 | |
| ≥97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only