missing translation for 'onlineSavingsMsg'
Learn More

(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl, 97+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6092103

Quantity 1g

 View more versions of this product

Catalog No. AAH6092103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1365531-82-3
698.78
JXFDRZQHVPTIMH-UHFFFAOYSA-N
87110704
C44H44O4P2
MFCD19443623
s-2,2'-bis di-p-tolylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine

Specifications

(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl
C44H44O4P2
MFCD19443623
s-2,2'-bis di-p-tolylphosphino-4,4',6,6'-tetramethoxybiphenyl, r-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine, s-4,4',6,6'-tetramethoxybiphenyl-2,2'-diyl bis di-p-tolylphosphine
698.78
698.77
1365531-82-3
1g
Air sensitive
JXFDRZQHVPTIMH-UHFFFAOYSA-N
87110704
≥97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions