missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, 98%

Catalog No. p-7051858
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4212002 0.5 g
AA4212004 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4212002 Supplier Thermo Scientific Chemicals Supplier No. 04212002
Only null left

Chiral catalyst ligand | CAS: 100165-88-6 | C48H40P2 | 678.796 g/mol

(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl is chiral ligand widely used in asymmetric synthesis. It also used as excellent catalysts for asymmetric hydrogenation of alkenes and some cyclic anhydrides. BINAP is used in organic synthesis for enantioselective transformations catalyzed by its complexes of ruthenium, rhodium, and palladium

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100165-88-6
Molecular Formula C48H40P2
Molecular Weight (g/mol) 678.796
MDL Number MFCD00269856
InChI Key IOPQYDKQISFMJI-UHFFFAOYSA-N
Synonym r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap
PubChem CID 4189906
IUPAC Name [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C

Specifications

Melting Point 255°C to 257°C
Quantity 0.5 g
Solubility Information Insoluble in water.
Formula Weight 678.79
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only