missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(tert-Butoxycarbonyl)-3-pyrrolidinol 98.0+%, TCI America™

Catalog No. B30551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30551G 1 g
B30555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30551G Supplier TCI America Supplier No. B30551G

Chemical Identifiers

CAS 101469-92-5
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.239
MDL Number MFCD01317839
InChI Key APCBTRDHCDOPNY-ZETCQYMHSA-N
Synonym s-1-n-boc-3-hydroxypyrrolidine, s-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, s-1-boc-3-hydroxypyrrolidine, s-+-1-boc-3-hydroxypyrrolidine, tert-butyl 3s-3-hydroxypyrrolidine-1-carboxylate, s-n-boc-3-pyrrolidinol, s-1-boc-3-pyrrolidinol, s-bochp, n-tert-butoxycarbonyl-s-+-3-pyrrolidinol, s-+-n-boc-3-pyrrolidinol
PubChem CID 854055
IUPAC Name tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)O

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 1 g
Formula Weight 187.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1-(tert-Butoxycarbonyl)-3-pyrrolidinol