missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylpropyl isocyanate, 95%

Catalog No. AAL2024603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2024603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2024603 Supplier Thermo Scientific Chemicals Supplier No. L2024603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 164033-12-9 | C10H11NO | 161.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 164033-12-9
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD05664090
InChI Key XGPXXSJFZSZULR-JTQLQIEISA-N
Synonym s---1-phenylpropyl isocyanate, s-1-phenylpropyl isocyanate, 1s-1-isocyanatopropyl benzene, benzene, 1-isocyanatopropyl-, s, 5-1-isocyanatopropyl benzene, s-1-isocyanatopropyl benzene, benzene, 1s-1-isocyanatopropyl
PubChem CID 7018275
IUPAC Name [(1S)-1-isocyanatopropyl]benzene
SMILES CCC(C1=CC=CC=C1)N=C=O

Specifications

Boiling Point 105°C (17 mmHg)
Refractive Index 1.509
Quantity 1 g
UN Number UN2206
Beilstein 6799586
Sensitivity Moisture sensitive
Optical Rotation −38.5° (c=1, Chloroform)
Formula Weight 161.2
Percent Purity 95%
Chemical Name or Material (S)-(-)-1-Phenylpropyl isocyanate
Hazard Category H302+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only