missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylethylamine 98.0+%, TCI America™

Catalog No. P079325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P079325ML 25 mL
P0793100ML 100 mL
P0793500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P079325ML Supplier TCI America Supplier No. P079325ML

Chemical Identifiers

CAS 2627-86-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00064406
InChI Key RQEUFEKYXDPUSK-ZETCQYMHSA-N
Synonym s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine
PubChem CID 75818
ChEBI CHEBI:35321
IUPAC Name (1S)-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Color Yellow
Boiling Point 84°C
Quantity 25 mL
UN Number 2922
Formula Weight 121.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-Phenylethylamine
EINECSNumber (3)-2692
RTECSNumber DP5775000
TSCA Yes