missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylethylamine, ChiPros 99+%, ee 99.5%

Catalog No. p-7047401
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1911806 5 g
AAL1911814 25 g
AAL1911822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL1911806 Supplier Thermo Scientific Chemicals Supplier No. L1911806
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 2627-86-3 | C8H11N | 121.183 g/mol

(S)-(-)-1-Phenylethylamine is utilized in a one-pot, multi-component synthesis of a highly substituted, chiral pyrrole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-Phenylethylamine is utilized in a one-pot, multi-component synthesis of a highly substituted, chiral pyrrole.

Solubility
Slightly miscible with water.

Notes
Keep away from sources of ignition. Incompatible with acids, acid chlorides, acid anhydrides and strong oxidizing agents.

Chemical Identifiers

CAS 2627-86-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00064406
InChI Key RQEUFEKYXDPUSK-ZETCQYMHSA-N
Synonym s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine
PubChem CID 75818
ChEBI CHEBI:35321
IUPAC Name (1S)-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Melting Point -10°C
Density 0.948
Boiling Point 187°C
Flash Point 71°C (159°F)
Odor Amine-like
Refractive Index 1.526
Quantity 5 g
UN Number UN2735
Beilstein 2204907
Sensitivity Air Sensitive
Merck Index 14,6026
Solubility Information Slightly miscible with water.
Optical Rotation −30° (c=10 in Ethanol)
Formula Weight 121.18
Percent Purity ≥99%
Assay ee 99.5%
Chemical Name or Material (S)-(-)-1-Phenylethylamine
Show More Show Less
Hazard Category H227-H302+H312-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 220-098-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only