missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylethanol, 97+%

Catalog No. AAB2118803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2118803 1 g
AAB2118806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2118803 Supplier Thermo Scientific Chemicals Supplier No. B2118803

CAS: 1445-91-6 | C8H10O | 122.17 g/mol

It finds its application in the synthesis of optically active products. Used for the synthesis of various pharmaceutical compounds. It can also be used as a chiral derivatizing agent for determining the absolute configuration of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis of optically active products. Used for the synthesis of various pharmaceutical compounds. It can also be used as a chiral derivatizing agent for determining the absolute configuration of secondary alcohols.

Solubility
Soluble in water at 20°C 20g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1445-91-6
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064264
InChI Key WAPNOHKVXSQRPX-ZETCQYMHSA-N
PubChem CID 443135
ChEBI CHEBI:16346
SMILES C[C@H](O)C1=CC=CC=C1

Specifications

Melting Point 9°C to 11°C
Density 1.012
Boiling Point 97°C to 99°C (20 mmHg)
Flash Point 85°C (185°F)
Odor Mild
Refractive Index 1.528
Quantity 1 g
UN Number UN2937
Beilstein 2039797
Synonym s-1-phenylethanol, s---1-phenylethanol, 1s-1-phenylethanol, 1s-1-phenylethan-1-ol, unii-2mic4qly2x, 1-phenylethanol, s, 2mic4qly2x, s-1-phenethyl alcohol, s-1-phenyl-1-ethanol, s-1-phenylethan-1-ol
Solubility Information Soluble in water at 20°C 20g/L. Soluble in alcohol.
Optical Rotation −45° (c=5 in Methanol)
IUPAC Name (1S)-1-phenylethanol
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material (S)-(-)-1-Phenylethanol
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H318-H302-H335-H227-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALPHA-METHYLBENZYL ALCOHOL, LIQUID
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only