missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 126°C |
| Quantity | 1 g |
| Formula Weight | 182.31 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | (S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine |
Safety and Handling
| TSCA | No |