missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine 97.0+%, TCI America™

Catalog No. M11831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11831G 1 g
M11835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11831G Supplier TCI America Supplier No. M11831G
Only null left

Chemical Identifiers

CAS 84466-85-3
Molecular Formula C11H22N2
Molecular Weight (g/mol) 182.311
MDL Number MFCD00077821
InChI Key VCVPSRADUBPOKJ-NSHDSACASA-N
Synonym (S)-(-)-1-[(1-Methyl-2-pyrrolidinyl)methyl]piperidine
PubChem CID 11126915
IUPAC Name 1-[[(2S)-1-methylpyrrolidin-2-yl]methyl]piperidine
SMILES CN1CCCC1CN2CCCCC2

Specifications

Color Yellow
Boiling Point 126°C
Quantity 1 g
Formula Weight 182.31
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine