missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Boc-3-(methylamino)piperidine, 96%

Catalog No. AAH6661806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6661806 5 g
AAH6661803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6661806 Supplier Thermo Scientific Chemicals Supplier No. H6661806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 912368-73-1 | C11H22N2O2 | 214.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 912368-73-1
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD09752964
InChI Key XRRRUOWSHGFPTI-VIFPVBQESA-N
Synonym s-1-n-boc-3-methylaminopiperidine, s-tert-butyl 3-methylamino piperidine-1-carboxylate, s-3-methylamino-piperidine-1-carboxylic acid tert-butyl ester, 1-n-boc-3-s-methylamino-piperidine, tert-butyl 3s-3-methylamino piperidine-1-carboxylate, pubchem14246, s-n-boc-3-methylamino piperidine, s-1-boc-3-methylamino piperidine
PubChem CID 25631268
IUPAC Name tert-butyl (3S)-3-(methylamino)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)NC

Specifications

Color Colorless to Yellow
Density 0.985 g/mL
Flash Point 110°C (230°F)
Refractive Index 1.46
Quantity 5 g
UN Number UN2810
Sensitivity Moisture sensitive
Formula Weight 214.3
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Boc-3-(methylamino)piperidine
Show More Show Less
Hazard Category H301-H319
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only