CAS RN 912368-73-1
IUPAC Name:
tert-butyl 3-(methylamino)piperidine-1-carboxylate
Synonyms:
tert-butyl 3-(methylamino)piperidine-1-carboxylate
Molecular Weight (g/mol):
214.31
Molecular Formula:
C11H22N2O2
InChi Key:
XRRRUOWSHGFPTI-UHFFFAOYNA-N
SMILES:
CNC1CCCN(C1)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
Promotions Available
(S)-(+)-1-Boc-3-(methylamino)piperidine, 96%
(S)-(+)-1-Boc-3-(methylamino)piperidine, 96%
CAS: 912368-73-1 Molecular Formula: C11H22N2O2 Molecular Weight (g/mol): 214.309 MDL Number: MFCD09752964 InChI Key: XRRRUOWSHGFPTI-VIFPVBQESA-N Synonym: s-1-n-boc-3-methylaminopiperidine,s-tert-butyl 3-methylamino piperidine-1-carboxylate,s-3-methylamino-piperidine-1-carboxylic acid tert-butyl ester,1-n-boc-3-s-methylamino-piperidine,tert-butyl 3s-3-methylamino piperidine-1-carboxylate,pubchem14246,s-n-boc-3-methylamino piperidine,s-1-boc-3-methylamino piperidine PubChem CID: 25631268 IUPAC Name: tert-butyl (3S)-3-(methylamino)piperidine-1-carboxylate SMILES: CC(C)(C)OC(=O)N1CCCC(C1)NC