Learn More
Chemical Identifiers
| CAS | 143900-44-1 |
|---|---|
| Molecular Formula | C10H19NO3 |
| Molecular Weight (g/mol) | 201.27 |
| MDL Number | MFCD04115307 |
| InChI Key | UIJXHKXIOCDSEB-UHFFFAOYNA-N |
| Synonym | s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine |
| PubChem CID | 1514399 |
| IUPAC Name | tert-butyl (3S)-3-hydroxypiperidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCCC(O)C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC436440050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 143900-44-1 | |
| 201.27 | |
| UIJXHKXIOCDSEB-UHFFFAOYNA-N | |
| 1514399 | |
| CC(C)(C)OC(=O)N1CCCC(O)C1 |
| C10H19NO3 | |
| MFCD04115307 | |
| s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine | |
| tert-butyl (3S)-3-hydroxypiperidine-1-carboxylate |
Specifications
| 143900-44-1 | |
| 100.0 | |
| Authentic | |
| C10H19NO3 | |
| + 9.3 | |
| UIJXHKXIOCDSEB-UHFFFAOYNA-N | |
| tert-butyl (3S)-3-hydroxypiperidine-1-carboxylate | |
| 1514399 | |
| 97% |
| 96.0 | |
| 110°C | |
| 96% min. (GC) | |
| MFCD04115307 | |
| s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine | |
| CC(C)(C)OC(=O)N1CCCC(O)C1 | |
| 201.27 | |
| 201.26 | |
| (S)-1-BOC-3-Hydroxypiperidine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
RUO – Research Use Only