missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-BOC-3-hydroxypiperidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 436440050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC436440050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

143900-44-1
201.27
UIJXHKXIOCDSEB-UHFFFAOYNA-N
1514399
CC(C)(C)OC(=O)N1CCCC(O)C1
C10H19NO3
MFCD04115307
s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine
tert-butyl (3S)-3-hydroxypiperidine-1-carboxylate

Specifications

(S)-1-BOC-3-Hydroxypiperidine
96.0
110°C
96% min. (GC)
C10H19NO3
5g
s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine
CC(C)(C)OC(=O)N1CCCC(O)C1
201.27
201.26
143900-44-1
100.0
Authentic
Glass bottle
MFCD04115307
+ 9.3
UIJXHKXIOCDSEB-UHFFFAOYNA-N
tert-butyl (3S)-3-hydroxypiperidine-1-carboxylate
1514399
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...