missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 114715-38-7 |
|---|---|
| Molecular Formula | C11H16N2 |
| Molecular Weight (g/mol) | 176.26 |
| InChI Key | HBVNLKQGRZPGRP-NSHDSACASA-N |
| Synonym | s-+-1-benzyl-3-aminopyrrolidine, 3s-1-benzylpyrrolidin-3-amine, 3s-+-1-benzyl-3-aminopyrrolidine, s-n-benzyl-3-aminopyrrolidine, s-3-amino-1-n-benzyl-pyrrolidine, s-1-benzylpyrrolidin-3-amine, s-1-benzyl-3-aminopyrrolidine, s-3-amino-1-benzylpyrrolidine, s-bap, s-3-amino-1-n-benzylpyrrolidine |
| PubChem CID | 1519353 |
| IUPAC Name | (3S)-1-benzylpyrrolidin-3-amine |
| SMILES | C1CN(CC1N)CC2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC437880010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC437880050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 114715-38-7 | |
| 176.26 | |
| s-+-1-benzyl-3-aminopyrrolidine, 3s-1-benzylpyrrolidin-3-amine, 3s-+-1-benzyl-3-aminopyrrolidine, s-n-benzyl-3-aminopyrrolidine, s-3-amino-1-n-benzyl-pyrrolidine, s-1-benzylpyrrolidin-3-amine, s-1-benzyl-3-aminopyrrolidine, s-3-amino-1-benzylpyrrolidine, s-bap, s-3-amino-1-n-benzylpyrrolidine | |
| (3S)-1-benzylpyrrolidin-3-amine |
| C11H16N2 | |
| HBVNLKQGRZPGRP-NSHDSACASA-N | |
| 1519353 | |
| C1CN(CC1N)CC2=CC=CC=C2 |
Specifications
| 114715-38-7 | |
| 100.0 | |
| 108°C to 115°C (5.0mmHg) | |
| Authentic | |
| C11H16N2 | |
| + 2 | |
| HBVNLKQGRZPGRP-NSHDSACASA-N | |
| (3S)-1-benzylpyrrolidin-3-amine | |
| 1519353 | |
| 99% |
| 98.5 | |
| 1.0200g/mL | |
| 140°C | |
| 98.5% min. (GC) | |
| 1.02 | |
| s-+-1-benzyl-3-aminopyrrolidine, 3s-1-benzylpyrrolidin-3-amine, 3s-+-1-benzyl-3-aminopyrrolidine, s-n-benzyl-3-aminopyrrolidine, s-3-amino-1-n-benzyl-pyrrolidine, s-1-benzylpyrrolidin-3-amine, s-1-benzyl-3-aminopyrrolidine, s-3-amino-1-benzylpyrrolidine, s-bap, s-3-amino-1-n-benzylpyrrolidine | |
| C1CN(CC1N)CC2=CC=CC=C2 | |
| 176.26 | |
| 176.26 | |
| (S)-(+)-1-Benzyl-3-aminopyrrolidine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
RUO – Research Use Only