missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1-Benzyl-3-aminopyrrolidine, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 437880010
| Quantity | 1g |
|---|---|
| Packaging | Glass bottle |
Chemical Identifiers
| 114715-38-7 | |
| 176.26 | |
| s-+-1-benzyl-3-aminopyrrolidine, 3s-1-benzylpyrrolidin-3-amine, 3s-+-1-benzyl-3-aminopyrrolidine, s-n-benzyl-3-aminopyrrolidine, s-3-amino-1-n-benzyl-pyrrolidine, s-1-benzylpyrrolidin-3-amine, s-1-benzyl-3-aminopyrrolidine, s-3-amino-1-benzylpyrrolidine, s-bap, s-3-amino-1-n-benzylpyrrolidine | |
| (3S)-1-benzylpyrrolidin-3-amine |
| C11H16N2 | |
| HBVNLKQGRZPGRP-NSHDSACASA-N | |
| 1519353 | |
| C1CN(CC1N)CC2=CC=CC=C2 |
Specifications
| (S)-(+)-1-Benzyl-3-aminopyrrolidine | |
| 98.5 | |
| 1.0200g/mL | |
| 140°C | |
| 98.5% min. (GC) | |
| C11H16N2 | |
| 1.02 | |
| s-+-1-benzyl-3-aminopyrrolidine, 3s-1-benzylpyrrolidin-3-amine, 3s-+-1-benzyl-3-aminopyrrolidine, s-n-benzyl-3-aminopyrrolidine, s-3-amino-1-n-benzyl-pyrrolidine, s-1-benzylpyrrolidin-3-amine, s-1-benzyl-3-aminopyrrolidine, s-3-amino-1-benzylpyrrolidine, s-bap, s-3-amino-1-n-benzylpyrrolidine | |
| C1CN(CC1N)CC2=CC=CC=C2 | |
| 176.26 | |
| 176.26 |
| 114715-38-7 | |
| 100.0 | |
| 108°C to 115°C (5.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1g | |
| + 2 | |
| HBVNLKQGRZPGRP-NSHDSACASA-N | |
| (3S)-1-benzylpyrrolidin-3-amine | |
| 1519353 | |
| 99% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
Promotions
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.