missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Amino-2-propanol 98.0+%, TCI America™

Catalog No. p-7117165
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
A09751G 1 g
A09755G 5 g
2 options

Catalog No. A09751G

Supplier: TCI America A09751G

Only null left

Chemical Identifiers

CAS 2799-17-9
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064429
InChI Key HXKKHQJGJAFBHI-UHFFFAOYNA-N
Synonym s-+-1-amino-2-propanol, s-1-aminopropan-2-ol, 2s-1-aminopropan-2-ol, +-isopropanolamine, s-1-amino-2-propanol, 2-propanol, 1-amino-, 2s, 1-amino-2-propanol #, 2-propanol, 1-amino-, s, ambotzfal1033
PubChem CID 7311736
IUPAC Name 1-aminopropan-2-ol
SMILES CC(O)CN

Specifications

Color White
Boiling Point 160°C
Quantity 1 g
UN Number 3263
Formula Weight 75.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (S)-(+)-1-Amino-2-propanol