accessibility menu, dialog, popup

UserName

CH3HOH2N

CAS RN 2799-17-9

IUPAC Name: 1-aminopropan-2-ol
Synonyms: 1-amino-2-propanol
Molecular Weight (g/mol): 75.11
Molecular Formula: C3H9NO
InChi Key: HXKKHQJGJAFBHI-UHFFFAOYNA-N
SMILES: CC(O)CN
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)