missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-1-(4-Methoxyphenyl)ethylamine 98.0+%, TCI America™
Supplier: TCI America M151225G
Specifications
| (S)-(-)-1-(4-Methoxyphenyl)ethylamine | |
| Yellow | |
| C9H13NO | |
| 25 g | |
| s---1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethanamine, s-1-4-methoxyphenyl ethylamine, s-1-4-methoxy-phenyl-ethylamine, 1s-1-4-methoxyphenyl ethanamine, s--1-4-methoxyphenyl ethylamine, s-p-1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethan-1-amine, 1s-1-4-methoxyphenyl ethan-1-amine | |
| CC(C1=CC=C(C=C1)OC)N | |
| 151.209 | |
| 151.21 | |
| Liquid |
| 41851-59-6 | |
| 11 (10g/L H2O soln. 20°C) | |
| MFCD00671660 | |
| 2735 | |
| JTDGKQNNPKXKII-ZETCQYMHSA-N | |
| (1S)-1-(4-methoxyphenyl)ethanamine | |
| 793467 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 41851-59-6 | |
| 151.209 | |
| JTDGKQNNPKXKII-ZETCQYMHSA-N | |
| 793467 | |
| CC(C1=CC=C(C=C1)OC)N |
| C9H13NO | |
| MFCD00671660 | |
| s---1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethanamine, s-1-4-methoxyphenyl ethylamine, s-1-4-methoxy-phenyl-ethylamine, 1s-1-4-methoxyphenyl ethanamine, s--1-4-methoxyphenyl ethylamine, s-p-1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethan-1-amine, 1s-1-4-methoxyphenyl ethan-1-amine | |
| (1S)-1-(4-methoxyphenyl)ethanamine |
Safety and Handling
TSCA : No