missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Methoxyphenyl)ethylamine 98.0+%, TCI America™

Catalog No. M15125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15125G 5 g
M151225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M15125G Supplier TCI America Supplier No. M15125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41851-59-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00671660
InChI Key JTDGKQNNPKXKII-ZETCQYMHSA-N
Synonym s---1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethanamine, s-1-4-methoxyphenyl ethylamine, s-1-4-methoxy-phenyl-ethylamine, 1s-1-4-methoxyphenyl ethanamine, s--1-4-methoxyphenyl ethylamine, s-p-1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethan-1-amine, 1s-1-4-methoxyphenyl ethan-1-amine
PubChem CID 793467
IUPAC Name (1S)-1-(4-methoxyphenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)OC)N

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-(4-Methoxyphenyl)ethylamine