Learn More
(S)-1,4-Diazabicyclo[4.3.0]nonane, 98+%
CAS: 93643-24-4 | C7H14N2 | 126.20 g/mol
Supplier: Thermo Scientific Chemicals H5229503
Description
(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| (S)-1,4-Diazabicyclo[4.3.0]nonane | |
| C7H14N2 | |
| 1 g | |
| s-octahydropyrrolo 1,2-a pyrazine, s-1,4-diazabicyclo 4.3.0 nonane, 8as-octahydropyrrolo 1,2-a pyrazine, 8as-octahydropyrrolo 1,2-a piperazine, s-octahydro-pyrrolo 1,2-a pyrazine, 6s-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8as, 8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine, pubchem20597, 6s-1,4-diazabicyclo 4,3,0 nonane | |
| FTTATHOUSOIFOQ-SSDOTTSWSA-N | |
| (8aR)-octahydropyrrolo[1,2-a]pyrazine | |
| 781249 | |
| ≥98% |
| 93643-24-4 | |
| MFCD03787926 | |
| UN3267 | |
| Slightly soluble in water. | |
| C1C[C@@H]2CNCCN2C1 | |
| 126.20 | |
| 126.2 |
Chemical Identifiers
| 93643-24-4 | |
| 126.20 | |
| FTTATHOUSOIFOQ-SSDOTTSWSA-N | |
| 781249 |
| C7H14N2 | |
| MFCD03787926 | |
| s-octahydropyrrolo 1,2-a pyrazine, s-1,4-diazabicyclo 4.3.0 nonane, 8as-octahydropyrrolo 1,2-a pyrazine, 8as-octahydropyrrolo 1,2-a piperazine, s-octahydro-pyrrolo 1,2-a pyrazine, 6s-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8as, 8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine, pubchem20597, 6s-1,4-diazabicyclo 4,3,0 nonane | |
| C1C[C@@H]2CNCCN2C1 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only